LIF466082914 Life Chemicals Inc.

1-(4-methylphenyl)prop-2-yn-1-ol

  • MDL number MFCD09032688
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart