LIF466082916 Life Chemicals Inc.

4-chloro-1-methylpyrazolo[3,4-d]pyrimidine

  • MDL number MFCD00661967
  • Cas Number : 23000-43-3
  • Molecular Weight : 168.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart