LIF466082918 Life Chemicals Inc.

1,3-dimethylimidazolidine-2,4-dione

  • MDL number MFCD00509320
  • Cas Number : 24039-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart