LIF491378771 Life Chemicals Inc.

2-(piperazin-1-yl)quinazolin-4(3H)-one dihydrochloride

  • MDL number MFCD29993819
  • Cas Number : 1881328-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart