LIF491378774 Life Chemicals Inc.

8-azaspiro[bicyclo[3.2.1]octane-3,5'-oxazolidin]-2'-one hydrochloride

  • MDL number MFCD30002044
  • Cas Number : 1949815-69-3
  • Molecular Weight : 218.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart