LIF491378781 Life Chemicals Inc.

3-amino-4-(tert-butyl)benzoic acid hydrochloride

  • MDL number MFCD29764127
  • Cas Number : 91246-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart