LIF491378792 Life Chemicals Inc.

2-amino-1-cyclobutylethan-1-one hydrochloride

  • MDL number MFCD29764130
  • Cas Number : 1881328-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart