LIF491378795 Life Chemicals Inc.

3-azabicyclo[3.1.0]hexan-6-amine dihydrochloride

  • MDL number MFCD08705927
  • Cas Number : 1369494-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart