LIF491378796 Life Chemicals Inc.

5-[(tert-butoxy)carbonyl]-4H,5H,6H,7H-thieno[3,2-c]pyridine-4-carboxylic acid

  • MDL number MFCD16529885
  • Cas Number : 1291217-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart