LIF491378798 Life Chemicals Inc.

7-(2-furyl)-1,4-thiazepan-5-one

  • MDL number MFCD24387727

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart