LIF491378801 Life Chemicals Inc.

1-methyl-3-[(prop-2-en-1-yl)amino]pyrrolidin-2-one

  • MDL number MFCD20284741
  • Cas Number : 1343805-37-7
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart