LIF491378804 Life Chemicals Inc.

4-(2-hydroxyethyl)-1,4-diazepane-1-carbaldehyde

  • MDL number MFCD18327985
  • Cas Number : 1342061-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart