LIF491378805 Life Chemicals Inc.

1-(cyclobutylmethyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD21241255
  • Cas Number : 1465204-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart