LIF491378813 Life Chemicals Inc.

8-(azetidin-3-yloxy)quinoline dihydrochloride

  • MDL number MFCD24849140
  • Cas Number : 1449117-70-7
  • Molecular Weight : 273.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart