LIF491378821 Life Chemicals Inc.

1-(4-fluorobenzyl)-2,5-dimethyl-1H-pyrrole

  • MDL number MFCD09034895
  • Cas Number : 908294-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart