LIF491378822 Life Chemicals Inc.

1-(4-fluoro-3-(trifluoromethyl)phenyl)-2,5-dimethyl-1H-pyrrole

  • MDL number MFCD19484824
  • Cas Number : 1284503-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart