LIF491378823 Life Chemicals Inc.

4-(tert-butyl)-3-nitrobenzoic acid

  • MDL number MFCD00100040
  • Cas Number : 59719-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart