LIF491378824 Life Chemicals Inc.

3-(4-fluorophenyl)-1,2-thiazole-5-carboxylic acid

  • MDL number MFCD21304963
  • Cas Number : 1486096-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart