LIF491378825 Life Chemicals Inc.

4-(tert-butyl)-3-hydroxybenzaldehyde

  • MDL number MFCD16999737
  • Cas Number : 532966-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart