LIF491378831 Life Chemicals Inc.

5-(aminomethyl)-1-methylpyrimidine-2,4(1H,3H)-dione hydrochloride

  • MDL number MFCD30004483
  • Cas Number : 2034157-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart