LIF491378834 Life Chemicals Inc.

4-(7-methylimidazo[1,2-a]pyrimidin-2-yl)aniline

  • MDL number MFCD28012690
  • Cas Number : 1163179-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart