LIF491378838 Life Chemicals Inc.

5-(diethylamino)furan-2-carbaldehyde

  • MDL number MFCD02614961
  • Cas Number : 22868-59-3
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart