LIF491378840 Life Chemicals Inc.

2-(cyclopropylamino)-2-(pyridin-3-yl)acetic acid

  • MDL number MFCD12048574
  • Cas Number : 1218388-85-2
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart