LIF491378844 Life Chemicals Inc.

1-(9,9-dimethyl-9H-fluoren-2-yl)ethan-1-one

  • MDL number MFCD27920910
  • Cas Number : 72322-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart