LIF491378854 Life Chemicals Inc.

2-methyl-octahydropyrrolo[3,4-c]pyrrole

  • MDL number MFCD09608113
  • Cas Number : 86732-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart