LIF491378863 Life Chemicals Inc.

5-cyclopropyl-1,2,4-thiadiazol-3-amine

  • MDL number MFCD20348043
  • Cas Number : 1342142-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart