LIF491378867 Life Chemicals Inc.

O-isopentylhydroxylamine hydrochloride

  • MDL number MFCD02249339
  • Cas Number : 51951-35-0
  • Molecular Weight : 139.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart