LIF491378868 Life Chemicals Inc.

1-(1-amino-2-methylpropan-2-yl)cyclobutan-1-ol

  • MDL number MFCD21805456
  • Cas Number : 1523159-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart