LIF491378873 Life Chemicals Inc.

3-chloro-5-cyclopropyl-1,2,4-thiadiazole

  • MDL number MFCD28151720
  • Cas Number : 1597081-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart