LIF491378875 Life Chemicals Inc.

4-chloro-N,6-dimethylpyrimidin-2-amine

  • MDL number MFCD08272046
  • Cas Number : 5738-15-8
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart