LIF491378894 Life Chemicals Inc.

1-(aminomethyl)-4-fluoro-2,3-dihydroinden-1-ol

  • MDL number MFCD11591022
  • Cas Number : 1082399-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart