LIF491378895 Life Chemicals Inc.

1-(aminomethyl)-3-methyl-2,3-dihydroinden-1-ol

  • MDL number MFCD16301770
  • Cas Number : 1226149-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart