LIF491378896 Life Chemicals Inc.

1-(aminomethyl)-3,3-dimethyl-2H-inden-1-ol

  • MDL number MFCD16303283
  • Cas Number : 1226174-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart