LIF491378897 Life Chemicals Inc.

1-(aminomethyl)-5-fluoro-2,3-dihydroinden-1-ol

  • MDL number MFCD27392616
  • Cas Number : 1547039-10-0
  • Molecular Weight : 181.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart