LIF491378902 Life Chemicals Inc.

methyl 4-amino-3-(4-fluorophenyl)-1,2-thiazole-5-carboxylate

  • MDL number MFCD19705600
  • Cas Number : 82424-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart