LIF509443218 Life Chemicals Inc.

Trans-4-methylcyclohexylamine

  • MDL number MFCD06411232
  • Cas Number : 2523-55-9
  • Molecular Weight : 113.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart