LIF509443265 Life Chemicals Inc.

methyl 3-oxopentanoate

  • MDL number MFCD00011705
  • Cas Number : 30414-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart