LIF509444084 Life Chemicals Inc.

Phenylhydrazine-4-sulfonic acid

  • MDL number MFCD00025097
  • Cas Number : 98-71-5
  • Molecular Weight : 188.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart