LIF509444278 Life Chemicals Inc.

6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid

  • MDL number MFCD00052030
  • Cas Number : 27372-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart