LIF509444412 Life Chemicals Inc.

1,3-dibromoimidazo[1,5-a]pyridine

  • MDL number MFCD08692530
  • Cas Number : 72315-45-8
  • Molecular Weight : 275.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart