LIF509444414 Life Chemicals Inc.

1-(6-amino-3-azabicyclo[3.1.0]hexan-3-yl)ethan-1-one hydrochloride

  • MDL number MFCD29045791
  • Cas Number : 1989672-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart