LIF509444426 Life Chemicals Inc.

3-bromooxan-2-one

  • MDL number MFCD13659268
  • Cas Number : 55974-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart