LIF509444445 Life Chemicals Inc.

1-bromo-3-tert-butyl-5-fluorobenzene

  • MDL number MFCD13193246
  • Cas Number : 1123172-38-2
  • Molecular Weight : 231.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart