LIF509444447 Life Chemicals Inc.

1,1-difluorospiro[2.5]octan-6-ol

  • MDL number MFCD28963076
  • Cas Number : 1781334-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart