LIF509444630 Life Chemicals Inc.

1H-benzo[c][1,2]thiazin-4(3H)-one 2,2-dioxide

  • MDL number MFCD18325052
  • Cas Number : 7117-28-4
  • Molecular Weight : 197.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart