LIF509444634 Life Chemicals Inc.

2-thia-5-azabicyclo[2.2.1]heptane hydrochloride

  • MDL number MFCD17976515
  • Cas Number : 1215499-34-5
  • Molecular Weight : 151.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart