LIF509444740 Life Chemicals Inc.

N-ethyl-2-methyl-6-(piperazin-1-yl)pyrimidin-4-amine

  • MDL number MFCD23990424
  • Cas Number : 1507884-58-3
  • Molecular Weight : 221.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart