LIF509444747 Life Chemicals Inc.

2-(pyrimidin-2-yloxy)acetic acid

  • MDL number MFCD11048462
  • Cas Number : 328084-05-5
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart