LIF509444786 Life Chemicals Inc.

6-(dimethylamino)-5-hydroxynicotinic acid

  • MDL number MFCD29999490
  • Cas Number : 1949815-86-4
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart