LIF509444787 Life Chemicals Inc.

5-hydroxy-2,6-dimethoxynicotinic acid

  • MDL number MFCD29999491
  • Cas Number : 1949815-77-3
  • Molecular Weight : 199.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart